MilliporeSigma
  • Rifaximin versus nonabsorbable disaccharides in the management of hepatic encephalopathy: a meta-analysis.
Rifaximin versus nonabsorbable disaccharides in the management of hepatic encephalopathy: a meta-analysis.
European journal of gastroenterology & hepatology (2008-12-03)
Qian Jiang, Xue-Hua Jiang, Ming-Hua Zheng, Liu-Ming Jiang, Yong-Ping Chen, Li Wang
ABSTRACT
To compare the positive and negative effects of rifaximin and nonabsorbable disaccharides in patients with hepatic encephalopathy. We used the method recommended by The Cochrane Collaboration to perform a meta-analysis of comparative randomized trials of rifaximin and nonabsorbable disaccharides. Seven randomized controlled trials were identified, but only five trials involving 264 patients met all the inclusion criteria. There was no significant difference between rifaximin and nonabsorbable disaccharides on improvement in patients with hepatic encephalopathy [relative risk (RR) 1.08; 95% confidence interval (CI), 0.85-1.38; P=0.53]. RR was 0.98 (95% CI: 0.85-1.13; P=0.74) for acute hepatic encephalopathy in 157 patients and 0.87 (95% CI: 0.40-1.88; P=0.72) for chronic hepatic encephalopathy in 96 patients, respectively. There was no significant difference between rifaximin and nonabsorbable disaccharides on diarrhea (RR=0.90; 95% CI: 0.17-4.70; P=0.90). However, a significant difference in favor of rifaximin on abdominal pain (RR=0.28; 95% CI: 0.08-0.95; P=0.04) was identified. Rifaximin is not superior to nonabsorbable disaccharides for acute or chronic hepatic encephalopathy in the long-term or short-term treatment except that it may be better tolerated. Further studies on larger populations are required to provide more sufficient evidence for assessment of the use of rifaximin.
Product Description
Rifaximin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Rifaximin
Supelco
Rifaximin, VETRANAL®, analytical standard
Rifaximin, European Pharmacopoeia (EP) Reference Standard