MilliporeSigma
The azadirachtin story.
Angewandte Chemie (International ed. in English) (2008-11-26)
Gemma E Veitch, Alistair Boyer, Steven V Ley
ABSTRACT
Azadirachtin has been the subject of intensive research within the scientific community ever since its isolation from the neem tree in 1968. There are now over 1000 publications relating to this natural product which cover all aspects of structural, biological and synthetic studies. Herein, we describe the worldwide synthesis efforts towards this fascinating molecule.
Sigma-Aldrich
Azadirachtin, ~95%