MilliporeSigma
  • Successful use of miltefosine and sodium stibogluconate, in combination, for the treatment of an HIV-positive patient with visceral leishmaniasis: a case report and brief review of the literature.
Successful use of miltefosine and sodium stibogluconate, in combination, for the treatment of an HIV-positive patient with visceral leishmaniasis: a case report and brief review of the literature.
Annals of tropical medicine and parasitology (2009-07-09)
P Collini, N Premchand, D Lockwood, J Greig
Sigma-Aldrich
Miltefosine, ≥98% (perchloric acid titration)