MilliporeSigma
  • Acrolein in wine: importance of 3-hydroxypropionaldehyde and derivatives in production and detection.
Acrolein in wine: importance of 3-hydroxypropionaldehyde and derivatives in production and detection.
Journal of agricultural and food chemistry (2010-03-03)
Rolene Bauer, Donald A Cowan, Andrew Crouch
ABSTRACT
Certain lactic acid bacteria strains belonging to the genus Lactobacillus have been implicated in the accumulation of 3-hydroxypropionaldehyde (3-HPA) during anaerobic glycerol fermentation. In aqueous solution 3-HPA undergoes reversible dimerization and hydration, resulting in an equilibrium state between different derivatives. Wine quality may be compromised by the presence of 3-HPA due to the potential for spontaneous conversion into acrolein under winemaking conditions. Acrolein is highly toxic and has been implicated in the development of bitterness in wine. Interconversion between 3-HPA derivatives and acrolein is a complex and highly dynamic process driven by hydration and dehydration reactions. Acrolein is furthermore highly reactive and its steady-state concentration in complex systems very low. As a result, analytical detection and quantification in solution is problematic. This paper reviews the biochemical and environmental conditions leading to accumulation of its precursor, 3-HPA. Recent advances in analytical detection are summarized, and the roles played by natural chemical derivatives are highlighted.