MilliporeSigma
  • Pharmacogenetic screening of carbamazepine-induced severe cutaneous allergic reactions.
Pharmacogenetic screening of carbamazepine-induced severe cutaneous allergic reactions.
Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia (2011-08-02)
Chaichon Locharernkul, Vorasuk Shotelersuk, Nattiya Hirankarn
ABSTRACT
Recent studies associated the HLA-B 1502 allele with carbamazepine (CBZ)-induced Stevens-Johnson syndrome (SJS)/toxic epidermal necrolysis (TEN) in patients from China, Thailand and Malaysia. No association has been found in patients from Europe or Japan. Linkage summary reports from East and South-east Asia predict a highly significant odds ratio (OR) of 84.75 (95% confidence interval [CI]=42.53-168.91; p=8.96×10[-15]) with sensitivity and negative predictive values of 92% and 98%, respectively. The higher prevalence of HLA-B 1502 allele among certain Asian populations (10-15%) compared to Caucasians (1-2%) may explain a 10-fold to 25-fold higher incidence of CBZ-SJS/TEN in patients from Asia. Screening for HLA-B 1502 before using CBZ can prevent SJS/TEN in certain populations, but screening may be less beneficial in populations with low HLA-B 1502 allele frequency and in patients exposed to CBZ for more than 2 months. A retrospective study demonstrated that the costs of HLA-B 1502 screening were less than those of SJS treatment. This article reviews possible benefits and concerns of HLA-B 1502 screening in clinical practice.
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Supelco
Carbamazepine, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®