MilliporeSigma
  • Will mineral trioxide aggregate replace calcium hydroxide in treating pulpal and periodontal healing complications subsequent to dental trauma? A review.
Will mineral trioxide aggregate replace calcium hydroxide in treating pulpal and periodontal healing complications subsequent to dental trauma? A review.
Dental traumatology : official publication of International Association for Dental Traumatology (2011-09-08)
Leif K Bakland, Jens O Andreasen
ABSTRACT
Mineral trioxide aggregate (MTA) has over the last two decades begun to take the place of calcium hydroxide (CH) in the treatment of a variety of pulpal and periodontal healing complications following dental trauma. These conditions include teeth with: (i) exposed pulps, (ii) immature roots and pulp necrosis, (iii) root fractures and pulp necrosis located in the coronal part of the pulps, and (iv) external infection-related (inflammatory) root resorption. The main reasons for replacing CH with MTA in these situations have generally been the delayed effect when using CH to induce hard tissues, the quality of such induced hard tissues, and finally the dentin weakening effect of CH, which in some instances lead to cervical root fractures in immature teeth. MTA appears, from a relatively few clinical studies, to overcome these shortcomings of CH. The lack of long-term clinical studies, however, may warrant a certain reservation in an unrestricted replacement of CH with MTA. A definite need for randomized clinical studies comparing CH and MTA in trauma healing situations is urgently needed.
Sigma-Aldrich
Calcium hydroxide, 99.995% trace metals basis
Sigma-Aldrich
Calcium hydroxide, CP
Sigma-Aldrich
Calcium hydroxide, ACS reagent, ≥95.0%
Sigma-Aldrich
Calcium hydroxide, puriss. p.a., reag. Ph. Eur., ≥96%