MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A rare case of propofol-induced liver injury during modified electroconvulsive therapy in an elderly woman.
A rare case of propofol-induced liver injury during modified electroconvulsive therapy in an elderly woman.
Internal medicine (Tokyo, Japan) (2013-04-03)
Akira Asai, Mai Yagi, Yusuke Tsuchimoto, Shinya Fukunishi, Atsushi Takeshita, Yasuhiro Tsuda, Akira Fukuda, Kazuhide Higuchi
ABSTRACT
A 75-year-old woman developed depression in 2010 and was treated with oral medications at our Department of Psychiatry. Since she showed no tendency toward improvement, she underwent modified electroconvulsive therapy (mECT). Later, she developed severe liver injury that was presumably induced by the propofol used for mECT. Propofol is an intravenous anesthetic agent that reportedly can be used relatively safely in the presence of liver dysfunction. We herein report the first case of propofol-induced liver injury definitively diagnosed based on positive drug lympocyte stimulation testing (DLST).
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Supelco
Propofol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Propofol, European Pharmacopoeia (EP) Reference Standard