MilliporeSigma
Acid-base and potassium homeostasis.
Seminars in nephrology (2013-08-21)
L Lee Hamm, Kathleen S Hering-Smith, Nazih L Nakhoul
ABSTRACT
Acid-base balance and potassium disorders are often clinically linked. Importantly, acid-base disorders alter potassium transport. In general, acidosis causes decreased K(+) secretion and increased reabsorption in the collecting duct. Alkalosis has the opposite effects, often leading to hypokalemia. Potassium disorders also influence acid-base homeostasis. Potassium depletion causes increased H(+) secretion, ammoniagenesis and H-K-ATPase activity. Hyperkalemia decreases ammoniagenesis and NH4(+) transport in the thick ascending limb. Some combined potassium and acid-base disorders involve indirect factors such as aldosterone, impaired renal function, volume depletion, and diarrhea. In summary, disorders of potassium and acid-base homeostasis are mechanistically linked and clinically important.
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis