MilliporeSigma
  • Review of tissue penetration and clinical efficacy of enoxacin in skin and skin structure infections and in osteomyelitis.
Review of tissue penetration and clinical efficacy of enoxacin in skin and skin structure infections and in osteomyelitis.
Clinical pharmacokinetics (1989-01-01)
G A Pankey
ABSTRACT
Enoxacin achieves a high penetration into skin tissue and blister fluid, reaching a maximum serum concentration (Cmax) of 3.7 mg/L at a time to reach maximum concentration (tmax) of 1.9 hours and a blister-fluid Cmax of 2.9 mg/L at a tmax of 3.7 hours after an oral dose of 600 mg. The half-life of enoxacin is 6.2 hours in serum and 7.2 hours in blister fluid. In a multicentre, open, non-comparative trial, clinical cure or improvement in skin or skin structure infections was achieved after oral administration of enoxacin 200 to 600 mg twice daily in 88% of 196 evaluable patients. Overall satisfactory bacteriological response was obtained in 76% of patients. In a multicentre, randomised, double-blind trial comparing oral enoxacin 400 mg twice daily with cephalexin 500 mg twice daily, satisfactory clinical outcome was achieved in 92% of 73 evaluable patients receiving enoxacin and in 99% of 72 evaluable patients receiving cephalexin. Furthermore, there was no statistically significant difference between the bacteriological efficacy of the 2 agents. In 3 single-centre trials, satisfactory clinical results were achieved in 75 to 100% of patients, and satisfactory bacteriological results occurred in 47 to 76% of patients after administration of oral enoxacin 400 mg twice daily for 7 to 14 days. In vitro uptake of enoxacin in bone leads to a concentration of 300 micrograms/g, with 83% being retained by bone after 3 washings with saline at pH 7.2. Clinical trials involving oral enoxacin in osteomyelitis are currently under way.