MilliporeSigma
Disopyramide-induced hypoglycaemia and increased serum insulin.
The New Zealand medical journal (1987-07-08)
M S Croxson, D W Shaw, P G Henley, H D Gabriel
ABSTRACT
A patient treated with disopyramide presented with hypoglycaemia, a raised serum insulin level and died of pneumonia. From these findings and a review of 10 case reports, we propose that disopyramide causes hypoglycaemia by stimulation of insulin release as described for the antimalarial drugs quinine and quinidine.
Product Description
Disopyramide, European Pharmacopoeia (EP) Reference Standard