MilliporeSigma
Maleic hydrazide: should the Delaney amendment apply to its use?
Journal of toxicology and environmental health (1977-05-01)
T J Haley
PMID325222
ABSTRACT
The chemistry, metabolism, toxicology, mutagenicity, and carcinogenicity of maleic hydrazide have been reviewed. There is little doubt that this chemical is a mutagen and a carcinogen in cell cultures and animals, but no evidence is available on human carcinogenicity regardless of population exposure in manufacturing, agriculture, and the food chain (i.e., potato chips). An epidemiology survey should be conducted to ascertain possible human carcinogenicity in these populations. A long-term ingestion experiment should be conducted in several animal species to establish whether maleic hydrazide is carcinogenic by this route. Biotransformation studies should be undertaken along with pharmacokinetic studies to obtain a better understanding of the chemical's metabolism and excretion. Such investigations would firmly establish whether the tolerance formaleic hydrazide should remain or whether the use of the compound should be banned under the Delaney Amendment.
Sigma-Aldrich
Maleic hydrazide, ReagentPlus®, 99%