MilliporeSigma
Flecainide acetate: critical care administration guidelines.
Heart & lung : the journal of critical care (1988-01-01)
S Senior, C Baessler, M Monzo
ABSTRACT
Flecainide may play an important role in the treatment of ventricular arrhythmias because of its convenient dosing regimen and high rate of efficacy in controlling ventricular ectopic activity. In stable patients with normal or mildly reduced left ventricular function, flecainide has proved to be promising. In those patients with severe left ventricular dysfunction, lethal arrhythmias, and preexisting conduction system disease, caution must be exercised.
Sigma-Aldrich
Flecainide acetate salt
Pricing and availability is not currently available.
Flecainide acetate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.