MilliporeSigma
[Tranquilizing activity of adamantane derivatives].
Farmakologiia i toksikologiia (1980-03-01)
V S Zalesov, B A Barteĭl, A L Fridman, N A Kolobov, V V Zalesov
ABSTRACT
It was established in experiments on mice and rats carried out according to conventional procedures that 13 adamantane derivatives have a greater tranquilizing activity than amantadine, 3-bromadamantoyl-1 diazomethane exerting a stronger effect than trioxazin. Two adamantane derivatives were found to have an anticonvulsant activity.
Sigma-Aldrich
Adamantane, ≥99%