MilliporeSigma
Emergency management of major hydrofluoric acid exposures.
Burns : journal of the International Society for Burn Injuries (1995-02-01)
R L Sheridan, C M Ryan, W C Quinby, J Blair, R G Tompkins, J F Burke
ABSTRACT
Exposures of as little as 2.5 per cent of the body surface to concentrated hydrofluoric acid (HF) may be fatal. Survival after major HF exposures is facilitated by aggressive emergency management which, we feel, includes wound irrigation, subeschar injection of calcium gluconate, monitored supplementation of serum calcium, and prompt wound excision carried out as an emergency procedure. The following case reports and literature review illustrate the important differences between exposures to concentrated HF of as little as 2 per cent of the body surface and smaller exposures to concentrated HF or exposure to dilute HF.
Sigma-Aldrich
Hydrofluoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥48%
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%