MilliporeSigma
Clinical applications of zolmitriptan (Zomig, 311C90).
Cephalalgia : an international journal of headache (1997-12-17)
R B Lipton, W F Stewart
ABSTRACT
Zolmitriptan (Zomig, formerly 311C90) is a novel, oral antimigraine drug that is consistently effective and well tolerated in the acute treatment of migraine headache and its associated symptoms. The purpose of this article is to review data available from pharmacological and clinical trials with zolmitriptan and to summarize the clinically relevant features that distinguish this agent. We will review the attributes desirable in a migraine drug and use this as a template for assessing zolmitriptan. Zolmitriptan provides a new oral treatment option for physicians which should help improve patient outcomes.
Sigma-Aldrich
Zolmitriptan, ≥98% (HPLC)