MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Entrapment and kinetic resolution of stabilized axial and equatorial conformers of spiro-β-lactams.
Entrapment and kinetic resolution of stabilized axial and equatorial conformers of spiro-β-lactams.
The Journal of organic chemistry (2011-06-15)
Naveen Anand, Bhahwal A Shah, Munish Kapoor, Rajinder Parshad, Rattan L Sharma, Maninder S Hundal, Ajay P S Pannu, Prasad V Bharatam, Subhash C Taneja
ABSTRACT
The facile synthesis of the stabilized axial and equatorial conformers of spiro-β-lactams was achieved via entrapment of cyclohexanone imines (Schiff bases) with acetoxyacetyl chloride in a [2 + 2]-cycloaddition reaction followed by their kinetic resolution. The immobilization of the racemic substrates on an inert solid support significantly reduced the reaction time and improved the enantioselectivity of conformers during kinetic resolution. The mechanism of the formation of the spiro-β-lactams was explored using B3LYP/6-31+G* level quantum chemical calculations.
Sigma-Aldrich
Acetoxyacetyl chloride, 97%