MilliporeSigma
  • Rearrangements concerted with fragmentation of cyclopropylmethoxychlorocarbene and cyclobutoxychlorocarbene in hydrocarbon solvents and Ar matrices.
Rearrangements concerted with fragmentation of cyclopropylmethoxychlorocarbene and cyclobutoxychlorocarbene in hydrocarbon solvents and Ar matrices.
Journal of the American Chemical Society (2004-07-09)
Robert A Moss, Ronald R Sauers, Fengmei Zheng, Xiaolin Fu, Thomas Bally, Alexander Maltsev
ABSTRACT
Fragmentations of cyclopropylmethoxychlorocarbene (6) and cyclobutoxychlorocarbene (10) lead to rearrangments that afford mixtures of cyclopropylmethyl chloride (7), cyclobutyl chloride (8), and 3-butenyl chloride (9). Isotopic substitution studies show that these rearrangments are accompanied by partial exchange of the methylene groups within 6 and 10. Surprisingly, these processes that are typical of carbocations persist in hydrocarbon solvents such as pentane and cyclohexane-d(12). Quantum chemical calculations reveal that the cis-conformers of the incipient oxychlorocarbenes C(4)H(7)OC(..)Cl decay to C(4)H(7)Cl + CO via transient hydrogen bonded C(4)H(7)(delta)(+)...Cl(delta-) complexes which possess significant ion pair character, even in the gas phase or in nonpolar solvents. In contrast to benzyloxychlorocarbene, no free radicals are formed upon generation or photolysis of 6 or 10 in Ar matrixes, although acid chlorides (the recombination products of these radical pairs) are observed. The IR spectra obtained in these experiments show the presence of several conformers of the two C(4)H(7)OC(..)Cl.