MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Synthesis of a low band gap polymer and its application in highly efficient polymer solar cells.
Synthesis of a low band gap polymer and its application in highly efficient polymer solar cells.
Journal of the American Chemical Society (2009-10-14)
Jianhui Hou, Hsiang-Yu Chen, Shaoqing Zhang, Ruby I Chen, Yang Yang, Yue Wu, Gang Li
ABSTRACT
HOMO level of the PBDTTT-based polymer was successfully reduced by introducing an ketone group in place of the ester group. The average PCE of the PBDTTT-based devices reached 6.3% with a champion PCE result of 6.58%. Due to its highly efficient photovoltaic performance and more feasible synthesis approach, PBDTTT-C has the potential to be successfully applied in the large-scale manufacturing of polymer solar cells.