MilliporeSigma
  • A mechanism of the irreversible inactivation of bovine pancreatic ribonuclease by diethylpyrocarbonate. A general reaction of diethylpyrocarbonate . A general reaction of diethylpyrocarbonate with proteins.
A mechanism of the irreversible inactivation of bovine pancreatic ribonuclease by diethylpyrocarbonate. A general reaction of diethylpyrocarbonate . A general reaction of diethylpyrocarbonate with proteins.
European journal of biochemistry (1970-04-01)
B Wolf, J A Lesnaw, M E Reichmann
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)