MilliporeSigma
  • Pegylated interferon β-1a for relapsing-remitting multiple sclerosis (ADVANCE): a randomised, phase 3, double-blind study.
Pegylated interferon β-1a for relapsing-remitting multiple sclerosis (ADVANCE): a randomised, phase 3, double-blind study.
The Lancet. Neurology (2014-05-06)
Peter A Calabresi, Bernd C Kieseier, Douglas L Arnold, Laura J Balcer, Alexey Boyko, Jean Pelletier, Shifang Liu, Ying Zhu, Ali Seddighzadeh, Serena Hung, Aaron Deykin
ABSTRACT
Subcutaneous pegylated interferon (peginterferon) beta-1a is being developed for treatment of relapsing multiple sclerosis, with less frequent dosing than currently available first-line injectable treatments. We assessed the safety and efficacy of peginterferon beta-1a after 48 weeks of treatment in the placebo-controlled phase of the ADVANCE trial, a study of patients with relapsing-remitting multiple sclerosis. We did this 2-year, double-blind, parallel group, phase 3 study, with a placebo-controlled design for the first 48 weeks, at 183 sites in 26 countries. Patients with relapsing-remitting multiple sclerosis (age 18-65 years, with Expanded Disability Status Scale score ≤5) were randomly assigned (1:1:1) via an interactive voice response or web system, and stratified by site, to placebo or subcutaneous peginterferon beta-1a 125 μg once every 2 weeks or every 4 weeks. The primary endpoint was annualised relapse rate at 48 weeks. This trial is registered with ClinicalTrials.gov, number NCT00906399. We screened 1936 patients and enrolled 1516, of whom 1512 were randomly assigned (500 to placebo, 512 to peginterferon every 2 weeks, 500 to peginterferon every 4 weeks); 1332 (88%) patients completed 48 weeks of treatment. Adjusted annualised relapse rates were 0·397 (95% CI 0·328-0·481) in the placebo group versus 0·256 (0·206-0·318) in the every 2 weeks group and 0·288 (0·234-0·355) in the every 4 weeks group (rate ratio for every 2 weeks group 0·644, 95% CI 0·500-0·831, p=0·0007; rate ratio for the every 4 weeks group 0·725, 95% CI 0·565-0·930, p=0·0114). 417 (83%) patients taking placebo, 481 (94%) patients taking peginterferon every 2 weeks, and 472 (94%) patients taking peginterferon every 4 weeks reported adverse events including relapses. The most common adverse events associated with peginterferon beta-1a were injection site reactions, influenza-like symptoms, pyrexia, and headache. 76 (15%) patients taking placebo, 55 (11%) patients taking study drug every 2 weeks, and 71 (14%) patients taking study drug every 4 weeks reported serious adverse events; relapse, pneumonia, and urinary tract infection were the most common. After 48 weeks, peginterferon beta-1a significantly reduced relapse rate compared with placebo. The drug might be an effective treatment for relapsing-remitting multiple sclerosis with less frequent administration than available treatments. Biogen Idec.
Product Description
Interferon β-1a, European Pharmacopoeia (EP) Reference Standard