MilliporeSigma
  • Design, synthesis, and pharmacological evaluation of a novel series of pyridopyrazine-1,6-dione γ-secretase modulators.
Design, synthesis, and pharmacological evaluation of a novel series of pyridopyrazine-1,6-dione γ-secretase modulators.
Journal of medicinal chemistry (2014-01-17)
Martin Pettersson, Douglas S Johnson, Chakrapani Subramanyam, Kelly R Bales, Christopher W am Ende, Benjamin A Fish, Michael E Green, Gregory W Kauffman, Patrick B Mullins, Thayalan Navaratnam, Subas M Sakya, Cory M Stiff, Tuan P Tran, Longfei Xie, Liming Zhang, Leslie R Pustilnik, Beth C Vetelino, Kathleen M Wood, Nikolay Pozdnyakov, Patrick R Verhoest, Christopher J O'Donnell
ABSTRACT
Herein we describe the design and synthesis of a novel series of γ-secretase modulators (GSMs) that incorporates a pyridopiperazine-1,6-dione ring system. To align improved potency with favorable ADME and in vitro safety, we applied prospective physicochemical property-driven design coupled with parallel medicinal chemistry techniques to arrive at a novel series containing a conformationally restricted core. Lead compound 51 exhibited good in vitro potency and ADME, which translated into a favorable in vivo pharmacokinetic profile. Furthermore, robust reduction of brain Aβ42 was observed in guinea pig at 30 mg/kg dosed orally. Through chemical biology efforts involving the design and synthesis of a clickable photoreactive probe, we demonstrated specific labeling of the presenilin N-terminal fragment (PS1-NTF) within the γ-secretase complex, thus gaining insight into the binding site of this series of GSMs.