MilliporeSigma
  • Solid-phase extraction of phosphorous-containing amino acid herbicides from biological specimens with a zirconia-coated silica cartridge
Solid-phase extraction of phosphorous-containing amino acid herbicides from biological specimens with a zirconia-coated silica cartridge
Journal of Chromatography. B, Biomedical Applications (2014)
Watanabe, D., et al.