MilliporeSigma
  • Reactions of phenanthraquinone and retenequinone with aldehydes and ammonium acetate in acetic acid solution1.
Reactions of phenanthraquinone and retenequinone with aldehydes and ammonium acetate in acetic acid solution1.
Journal of the American Chemical Society (1943)
Steck EA and Day AR.