MilliporeSigma
  • Characterization of novel stem cell factor responsive human mast cell lines LAD 1 and 2 established from a patient with mast cell sarcoma/leukemia; activation following aggregation of FcepsilonRI or FcgammaRI.
Characterization of novel stem cell factor responsive human mast cell lines LAD 1 and 2 established from a patient with mast cell sarcoma/leukemia; activation following aggregation of FcepsilonRI or FcgammaRI.
Leukemia research (2003-06-13)
Arnold S Kirshenbaum, Cem Akin, Yalin Wu, Menachem Rottem, Julie P Goff, Michael A Beaven, V Koneti Rao, Dean D Metcalfe
ABSTRACT
Two novel stem cell factor (SCF) dependent human mast cell lines, designated LAD 1 and 2, were established from bone marrow aspirates from a patient with mast cell sarcoma/leukemia. LAD 1 and 2 cells have the ultrastructural features of human mast cells, and express FcepsilonRI, CD4, 9, 13, 14, 22, 31, 32, 45, 64, 71, 103, 117, 132, CXCR4 (CD184), CCR5 (CD195); and intracytoplasmic histamine, tryptase and chymase. LAD 1 and 2 do not exhibit activating mutations at codon 816 of c-kit. Both LAD 1 and 2 release beta-hexosaminidase following FcepsilonRI or FcgammaRI aggregation. The availability of these cell lines offers an unparalleled circumstance to examine the biology of human mast cells.
Sigma-Aldrich
pZerve, Cryopreservation Solution
Pricing and availability is not currently available.