MilliporeSigma
  • Comparison of two methods for detecting varicella-zoster virus antibody with varicella-zoster virus cell-mediated immunity.
Comparison of two methods for detecting varicella-zoster virus antibody with varicella-zoster virus cell-mediated immunity.
Journal of clinical microbiology (1996-02-01)
A Weinberg, A R Hayward, H B Masters, I A Ogu, M J Levin
ABSTRACT
We evaluated an enzyme-linked immunoassay (EIA; BioWhittaker) and a latex agglutination (LA; Becton Dickinson) for varicella-zoster virus (VZV) antibody determination, using cell-mediated immunity (CMI) as a "gold standard." VZV EIA had a sensitivity, specificity, positive predictive value, and negative predictive value of 87, 91, 87, and 91%, respectively, compared with CMI. Correlation was excellent except when the varicella index was 0.9 to 1.2. We defined sera with varicella indices of 0.9 to 1.2 as indeterminate. LA had a sensitivity, specificity, positive predictive value, and negative predictive value of 96, 91, 97, and 90%, respectively, compared with EIA. LA reactivity only at a 1:2 dilution did not correlate with CMI, but sera reactive at dilutions of > or = 1:8 indirectly did. We defined indeterminate sera as those reactive at 1:2 and nonreactive at 1:8. EIA and LA were equivalent for determining VZV immune status, and both methods required modified criteria of interpretation to increase their specificity.