MilliporeSigma
Caspases: killer proteases.
Trends in biochemical sciences (1997-08-01)
D W Nicholson, N A Thornberry
ABSTRACT
Caspases (cysteinyl aspartate-specific proteinases) mediate highly specific proteolytic cleavage events in dying cells, which collectively manifest the apoptotic phenotype. The key and central role that these enzymes play in a biochemical cell-suicide pathway has been conserved throughout the evolution of multicellular eukaryotes.