MilliporeSigma
2,4,6-Trifluoro-benzoic acid.
Acta crystallographica. Section E, Structure reports online (2011-04-28)
Richard Betz, Thomas Gerber
ABSTRACT
In the title compound, C(7)H(3)F(3)O(2), the C-C-C angles in the ring are greater than 120° for F-bonded C atoms [123.69 (13), 123.88 (12) and 123.66 (12)°]. In the crystal, inter-molecular O-H⋯O hydrogen bonds between carboxyl groups give rise to the formation of a centrosymmetric dimer, while dispersive F⋯O contacts [2.8849 (16) Å] connect the dimers into infinite strands along the a axis.