MilliporeSigma
  • GEP100 links epidermal growth factor receptor signalling to Arf6 activation to induce breast cancer invasion.
GEP100 links epidermal growth factor receptor signalling to Arf6 activation to induce breast cancer invasion.
Nature cell biology (2007-12-18)
Masaki Morishige, Shigeru Hashimoto, Eiji Ogawa, Yoshinobu Toda, Hirokazu Kotani, Mayumi Hirose, Shumei Wei, Ari Hashimoto, Atsuko Yamada, Hajime Yano, Yuichi Mazaki, Hiroshi Kodama, Yoshinori Nio, Toshiaki Manabe, Hiromi Wada, Hidenori Kobayashi, Hisataka Sabe
ABSTRACT
Epidermal growth factor (EGF) receptor (EGFR) signalling is implicated in tumour invasion and metastasis. However, whether there are EGFR signalling pathways specifically used for tumour invasion still remains elusive. Overexpression of Arf6 and its effector, AMAP1, correlates with and is crucial for the invasive phenotypes of different breast cancer cells. Here we identify the mechanism by which Arf6 is activated to induce tumour invasion. We found that GEP100/BRAG2, a guanine nucleotide exchanging factor (GEF) for Arf6, is responsible for the invasive activity of MDA-MB-231 breast cancer cells, whereas the other ArfGEFs are not. GEP100, through its pleckstrin homology domain, bound directly to Tyr1068/1086-phosphorylated EGFR to activate Arf6. Overexpression of GEP100, together with Arf6, caused non-invasive MCF7 cells to become invasive, which was dependent on EGF stimulation. Moreover, GEP100 knockdown blocked tumour metastasis. GEP100 was expressed in 70% of primary breast ductal carcinomas, and was preferentially co-expressed with EGFR in the malignant cases. Our results indicate that GEP100 links EGFR signalling to Arf6 activation to induce invasive activities of some breast cancer cells, and hence may contribute to their metastasis and malignancy.