MilliporeSigma
  • Pancreatic lipase-related protein 2 digests fats in human milk and formula in concert with gastric lipase and carboxyl ester lipase.
Pancreatic lipase-related protein 2 digests fats in human milk and formula in concert with gastric lipase and carboxyl ester lipase.
Pediatric research (2013-06-05)
Karin Johnson, Leah Ross, Rita Miller, Xunjun Xiao, Mark E Lowe
ABSTRACT
Dietary fats must be digested into fatty acids and monoacylglycerols prior to absorption. In adults, colipase-dependent pancreatic triglyceride lipase (PTL) contributes significantly to fat digestion. In newborn rodents and humans, the pancreas expresses low levels of PTL. In rodents, a homologue of PTL, pancreatic lipase-related protein 2 (PLRP2), and carboxyl ester lipase (CEL) compensate for the lack of PTL. In human newborns, the role of PLRP2 in dietary fat digestion is unclear. To clarify the potential of human PLRP2 to influence dietary fat digestion in newborns, we determined PLRP2 activity against human milk and infant formula. The activity of purified recombinant PLRP2, gastric lipase (GL), and CEL against fats in human milk and formula was measured with each lipase alone and in combination with a standard pH-stat assay. Colipase added to human milk stimulated fat digestion. PLRP2 and CEL had activity against human milk and formula. Predigestion with GL increased PLRP2 activity against both substrates. Together, CEL and PLRP2 activity was additive with formula and synergistic with human milk. PLRP2 can digest fats in human milk and formula. PLRP2 acts in concert with CEL and GL to digest fats in human milk in vitro.