MilliporeSigma
Granulomatous dacryoadenitis in regional enteritis (crohn disease).
American journal of ophthalmology (2014-07-19)
Frederick A Jakobiec, Alia Rashid, Katherine A Lane, Michael Kazim
ABSTRACT
To evaluate the clinical and immunopathologic features of 2 patients with bilateral dacryoadenitis associated with regional enteritis. Retrospective, clinicopathologic study. Clinical records, photographs, and imaging studies were reviewed and microscopic sections of lacrimal gland biopsy samples were critically re-evaluated. The microscopic slides were stained with hematoxylin and eosin, special stains for organisms, and a range of immunohistochemical biomarkers, including CD3, CD4, CD5, CD8, CD20, CD68, CD138, CD1a, and immunoglobulins Ig G, IgG4, and IgA. Both patients were young women with a well-established diagnosis of regional enteritis. Histopathologic examination of biopsy samples disclosed moderate intraparenchymal fibrosis and lymphoplasmacytic infiltrates without lymphoid follicles. Small to medium intraparenchymal, noncaseating granulomas lacking multinucleated giant cells and, in 1 patient, CD68-positive and CD1a-negative palisading granulomas in widened interlobular fibrous septa were detected. Vasculitis and IgG4 plasma cells were not observed. Additional immunohistochemical studies revealed that CD8 T lymphocytes (suppressor or cytotoxic subset) predominated over CD4-positive T lymphocytes (helper cells) surrounding the necrobiotic foci and were intermixed with the CD68-positive histiocytes in the absence of CD20 B lymphocytes. Special stains for organisms demonstrated negative results. Dacryoadenitis is the rarest form of ocular adnexal involvement in regional enteritis, which affects the orbit far more frequently than ulcerative colitis. It is a granulomatous process with the possibility of palisading necrobiotic foci. In contrast, ulcerative colitis causes an interstitial lymphocytic and nongranulomatous myositis. Sarcoidosis, Wegener granulomatosis, and pseudorheumatoid nodules must be ruled out. Treatment options entail a wide variety of agents with selection based on empirical considerations and tailored to the patient's symptoms.
Sigma-Aldrich
Sulfasalazine, 97.0-101.5%