MilliporeSigma
  • Impact of UV-B on drought- or cadmium-induced changes in the fatty acid composition of membrane lipid fractions in wheat.
Impact of UV-B on drought- or cadmium-induced changes in the fatty acid composition of membrane lipid fractions in wheat.
Ecotoxicology and environmental safety (2014-07-26)
Orsolya Kinga Gondor, Gabriella Szalai, Viktória Kovács, Tibor Janda, Magda Pál
ABSTRACT
UV-B radiation may have either a positive or negative impact under the same conditions in wheat, depending on the type of secondary abiotic stressor: Cd or drought. Supplemental UV-B prevented the wilting and leaf rolling induced by PEG treatment. In contrast, combined UV-B and Cd treatment resulted in pronounced oxidative stress. The opposite effect of UV-B radiation in the case of drought or cadmium stress may be related to the alteration induced in the fatty acid composition. UV-B caused changes in the unsaturation of leaf phosphatidylglycerol fractions, and the accumulation of flavonoid in the leaves may prevent the stress induced by subsequent drought treatment. However it resulted in pronounced injury despite the increased flavonoid content in roots exposed to Cd. This was manifested in a drastic decrease in the unsaturation of the leaf monogalactosyldiacylglycerol and the root phosphatidylglycerol and digalactosyldiacylglycerol fractions. Data on the flavonoid content and fatty acid composition showed that oxidative stress was induced by drought in the leaves, by Cd in the roots, and interestingly, by UV-B radiation in both the leaves and roots. The additive effect of the combined stresses was also detected in the roots. The results presented here suggest a relationship between the capacity of the plant to remodel the fatty acid composition and its resistance to various stress factors.
Sigma-Aldrich
Distilled water, suitable for HPLC
Supelco
Water
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, wire reel, 0.1m, diameter 1.0mm, hard, 99.999%
Cadmium, foil, 15mm disks, thickness 0.125mm, as rolled, 99.95%