MilliporeSigma
  • Expression of four CEA family antigens (CEA, NCA, BGP and CGM2) in normal and cancerous gastric epithelial cells: up-regulation of BGP and CGM2 in carcinomas.
Expression of four CEA family antigens (CEA, NCA, BGP and CGM2) in normal and cancerous gastric epithelial cells: up-regulation of BGP and CGM2 in carcinomas.
International journal of cancer (1998-04-09)
T Kinugasa, M Kuroki, H Takeo, Y Matsuo, K Ohshima, Y Yamashita, T Shirakusa, Y Matsuoka
ABSTRACT
Four human carcinoembryonic antigen (CEA) family members, CEA (CD66e), non-specific cross-reacting antigen (NCA, CD66c), biliary glycoprotein (BGP, CD66a) and CEA gene-family member 2 (CGM2), are expressed in normal mucosal epithelia of the colon. Expression of BGP and CGM2 has recently been demonstrated to be down-regulated in colorectal adenocarcinomas. We have now investigated the expression of the 4 CEA family antigens in gastric adenocarcinoma and carcinoma cell lines in comparison with adjacent normal gastric mucosa. The transcripts of the CEA, NCA and BGP genes evaluated by reverse transcription-polymerase chain reaction were detectable at various levels in all the gastric adenocarcinoma cell lines tested, while CGM2 mRNA was detectable in the cell lines of poorly differentiated but not of well-differentiated carcinomas. The levels of CEA mRNA in normal gastric mucosa were variable but mostly increased in adenocarcinomas. The sparse expression of NCA observed in the normal tissues was markedly up-regulated in the carcinomas. In contrast to previous findings on normal and cancerous colonic tissues, the transcripts of CGM2 were totally undetectable and those of BGP were recognized only marginally, if at all, in normal gastric mucosa, while both messages were detected at significant levels in most of the gastric adenocarcinomas. This was confirmed by in situ hybridization. Our findings indicate that expression of the CEA family antigens, particularly that of BGP and CGM2, is differently regulated in epithelial cells of the colon and the stomach.