MilliporeSigma
  • Highly efficient and selective transport of methylene blue through a bulk liquid membrane containing Cyanex 301 as carrier.
Highly efficient and selective transport of methylene blue through a bulk liquid membrane containing Cyanex 301 as carrier.
Separation and Purification Technology (2011)
Madaeni SS, et al.