MilliporeSigma
  • Cyclopolymerization. IV. Structure of polymethacrylic anhydride and kinetics of polymerization of methacrylic anhydride.
Cyclopolymerization. IV. Structure of polymethacrylic anhydride and kinetics of polymerization of methacrylic anhydride.
Journal of Polymer Science: Part A, General Papers (1964)
Smets G, et al.