MilliporeSigma
  • Role of the putative tumor metastasis suppressor gene Drg-1 in breast cancer progression.
Role of the putative tumor metastasis suppressor gene Drg-1 in breast cancer progression.
Oncogene (2004-06-09)
Sucharita Bandyopadhyay, Sudha K Pai, Shigeru Hirota, Sadahiro Hosobe, Yukio Takano, Ken Saito, David Piquemal, Therese Commes, Misako Watabe, Steven C Gross, Ying Wang, Sophia Ran, Kounosuke Watabe
ABSTRACT
The differentiation-related gene-1 (Drg-1) was first identified as a gene strongly upregulated by induction of differentiation in colon carcinoma cells in vitro, and later the same gene was shown to suppress tumorigenicity of human bladder cancer cells in vivo. On the other hand, we and others have demonstrated that the Drg-1 gene suppresses prostate and colon cancer metastases in mouse models. In the context of such potential organ-specific differential function of the Drg-1 gene, the present study was designed to clarify the expression status, regulation and function of Drg-1 in the case of human breast cancer. We found that the expression of the Drg-1 protein was significantly reduced in breast tumor cells, particularly in patients with lymph node or bone metastasis as compared to those with localized breast cancer. Drg-1 expression also exhibited significant inverse correlation with the disease-free survival rate of patients and emerged as an independent prognostic factor. The downregulation of the Drg-1 gene appeared to be largely at the RNA level, and the DNA methylation inhibitor, 5-Azacytidine, significantly elevated the Drg-1 gene expression in various breast tumor cell lines. Furthermore, we found that overexpression of the Drg-1 gene suppresses the invasiveness of breast cancer cells in vitro, and this suppression was also achieved by treatment of cells with 5-Azacytidine. Together, our results strongly suggest functional involvement of the Drg-1 gene in suppressing the metastatic advancement of human breast cancer.