MilliporeSigma
  • Ligand-dependent interaction of the aryl hydrocarbon receptor with a novel immunophilin homolog in vivo.
Ligand-dependent interaction of the aryl hydrocarbon receptor with a novel immunophilin homolog in vivo.
The Journal of biological chemistry (1997-04-25)
L A Carver, C A Bradfield
ABSTRACT
In an effort to identify regulators of aryl hydrocarbon receptor (AHR) signaling, we have employed the yeast two-hybrid system to screen for human proteins that interact in a ligand-dependent manner with the AHR. After screening 1.4 x 10(6) clones from a human B cell library, two distinct clones were identified that associated specifically with the liganded receptor. No clones were identified that interacted preferentially with the unliganded AHR. One of the ligand-dependent clones, ARA9, encodes a novel 330-amino acid protein with regions of amino acid sequence similarity to the 52-kDa FK506-binding protein known to be associated with the glucocorticoid receptor. Yeast two-hybrid experiments with ARA9 demonstrated a strong interaction with the AHR that is enhanced 11-fold in the presence of the ligand beta-naphthoflavone. In vitro experiments using proteins generated in reticulocyte lysates confirmed this interaction and indicated that ARA9 can be co-immunoprecipitated with the AHR using antisera raised specifically for either the AHR or the 90-kDa heat shock protein. The observation that ARA9 has a high affinity for both the 90-kDa heat shock protein-associated and ligand-activated forms of the AHR suggests that ARA9 is a component of the AHR-signaling pathway in vivo.