MilliporeSigma
  • Peripheral nervous system plasmalogens regulate Schwann cell differentiation and myelination.
Peripheral nervous system plasmalogens regulate Schwann cell differentiation and myelination.
The Journal of Clinical Investigation (2014)
da Silva TF, Eira J, Lopes AT, et al.