MilliporeSigma
Unprecedented role of hydronaphthoquinone tautomers in biosynthesis.
Angewandte Chemie (International ed. in English) (2014-07-23)
Syed Masood Husain, Michael A Schätzle, Steffen Lüdeke, Michael Müller
ABSTRACT
Quinones and hydroquinones are among the most common cellular cofactors, redox mediators, and natural products. Here, we report on the reduction of 2-hydroxynaphthoquinones to the stable 1,4-diketo tautomeric form of hydronaphthoquinones and their further reduction by fungal tetrahydroxynaphthalene reductase. The very high diastereomeric and enantiomeric excess, together with the high yield of cis-3,4-dihydroxy-1-tetralone, exclude an intermediary hydronaphthoquinone. Labeling experiments with NADPH and NADPD corroborated the formation of an unexpected 1,4-diketo tautomeric form of 2-hydroxyhydronaphthoquinone as a stable intermediate. Similar 1,4-diketo tautomers of hydronaphthoquinones were established as products of the NADPH-dependent enzymatic reduction of other 1,4-naphthoquinones, and as substrates for different members of the superfamily of short-chain dehydrogenases. We propose an essential role of hydroquinone diketo tautomers in biosynthesis and detoxification processes.