MilliporeSigma
  • Experimental protein mixture for validating tandem mass spectral analysis.
Experimental protein mixture for validating tandem mass spectral analysis.
Omics : a journal of integrative biology (2002-07-30)
Andrew Keller, Samuel Purvine, Alexey I Nesvizhskii, Sergey Stolyar, David R Goodlett, Eugene Kolker
ABSTRACT
Several methods have been used to identify peptides that correspond to tandem mass spectra. In this work, we describe a data set of low energy tandem mass spectra generated from a control mixture of known protein components that can be used to evaluate the accuracy of these methods. As an example, these spectra were searched by the SEQUEST application against a human peptide sequence database. The numbers of resulting correct and incorrect peptide assignments were then determined. We show how the sensitivity and error rate are affected by the use of various filtering criteria based upon SEQUEST scores and the number of tryptic termini of assigned peptides.
Sigma-Aldrich
α-Lactalbumin from bovine milk, Type III, calcium depleted, ≥85% (PAGE), lyophilized powder
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis