MilliporeSigma
The clinical efficacy of poly (ethylene glycol)-modified proteins.
Journal of Controlled Release : Official Journal of the Controlled Release Society (1990)
Fuertges F and Abuchowski A.