MilliporeSigma
  • Marker expression in peripheral T-cell lymphoma: a proposed clinical-pathologic prognostic score.
Marker expression in peripheral T-cell lymphoma: a proposed clinical-pathologic prognostic score.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2006-04-26)
Philip Went, Claudio Agostinelli, Andrea Gallamini, Pier Paolo Piccaluga, Stefano Ascani, Elena Sabattini, Francesco Bacci, Brunangelo Falini, Teresio Motta, Marco Paulli, Tullio Artusi, Milena Piccioli, Pier Luigi Zinzani, Stefano A Pileri
ABSTRACT
Although peripheral T-cell lymphoma, unspecified (PTCL/U), is the most common T-cell tumor in Western countries, no study to date has been based on the application of a wide panel of markers to a large series of patients and assessed the impact of phenotype on survival. We evaluated the expression of 19 markers in 148 PTCLs/U and 45 PTCLs of the angioimmunoblastic type (AILD). The analysis was performed on tissue microarrays by immunohistochemistry and in situ hybridization. Clinical data were available in 93 PTCL/U patients, most of whom had been included in a previous study proposing a prognostic index (PIT). An aberrant phenotype with frequent loss of CD5 and/or CD7 was typical for PTCLs, irrespective of whether they were U or AILD. Aberrantly expressed proteins rarely included CD20, CD15, and CD30. Positivity for Epstein-Barr virus-associated small RNAs and CD15 expression emerged as adverse prognostic factors. Among PTCLs/U, the proliferation-associated protein Ki-67 turned out to be prognostically relevant and was integrated in a new predictive score, incorporating age (> 60 years), high lactate dehydrogenase, poor performance status, and Ki-67 > or = 80%. This score was associated with the patient outcome (P < .0001) and was found to be more robust than PIT (P = .0043) in the present series. Our retrospective analysis shows a wide range of protein expression in PTCLs and proposes a new prognostic index. The latter represents one of the first examples of mixed score (including patient- and tumor-specific factors) applied to malignant lymphomas and may be the basis for future prospective therapeutic trials.
Sigma-Aldrich
CD7 (MRQ-56) Mouse Monoclonal Antibody
Sigma-Aldrich
CD7 (EP132) Rabbit Monoclonal Primary Antibody