MilliporeSigma
  • The utility of basal cell-specific anti-cytokeratin antibody (34 beta E12) in the diagnosis of prostate cancer. A review of 228 cases.
The utility of basal cell-specific anti-cytokeratin antibody (34 beta E12) in the diagnosis of prostate cancer. A review of 228 cases.
The American journal of surgical pathology (1995-03-01)
K J Wojno, J I Epstein
ABSTRACT
Basal cell-specific anti-cytokeratin antibody (34 beta E12) decorates the basal cells of benign prostatic epithelium by standard immunohistochemical techniques, whereas adenocarcinoma of the prostate lacks immunoreactivity with this antibody. We reviewed our experience with this antibody to determine its utility in the diagnosis of adenocarcinoma of the prostate as well as its pattern of usage at a tertiary medical center. In all, 7,242 prostate specimens from 5,262 men were seen at Johns Hopkins Hospital between 1/89 and 4/93. Immunostaining for basal cell-specific cytokeratin (34 beta E12) was originally used for diagnostic purposes in 289 questionable area from 228 cases; 45% of these cases were seen in consultation. The distribution of cases using 34 beta E12 was 52% needle biopsies, 32% transurethral prostatic resections (TURPs). 13% radical prostatectomies, and 3% open enucleations. These procedures constituted 2.8% of all needle biopsies, 7.2% of all TURPs, 1.7% of all radical prostatectomies, and 3.5% of all enucleations seen during this time period. For this study the hematoxylin and eosin stain was reviewed without knowledge of the original diagnosis, a diagnosis was favored, the 34 beta E12 stain was examined, and a final diagnosis was determined. The 34 beta E12 stain established (14%), confirmed (58%), or changed (2%) our favored diagnoses, while 18% remained or became equivocal. The 34 beta E12 stain was of no use in 8% of the cases, yet we felt we were still able to render a final diagnosis even without the help of the stain. The differential diagnoses in the questionable foci using 34 beta E12 were cancer versus focus of atypical glands (44%), adenosis (39%), prostatic intraepithelial neoplasia (PIN) (8%), basal cell hyperplasia (5%), and atrophy (4%). However, 34 beta E12 was used in only 15-20% of all cases of adenosis and basal cell hyperplasia and in < 2% of PIN and atrophy cases seen during this time. Reasons for equivocal results were loss of suspicious glands on cut downs used for staining (49%), too few glands to rely on negative staining (23%), technical problems (15%), limited number of positive staining glands in a small focus (7%), and cautery artifact (6%). Although equivocal cases tended to have fewer negative stained glands than cases diagnosed with cancer, there was no minimum number of negative stained glands required to establish a diagnosis of cancer. From these data we conclude that 34 beta E12 staining is a useful tool in confirming, establishing, or changing the diagnosis in questionable foci seen in the everyday practice of surgical pathology.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
Cytokeratin (34betaE12) Mouse Monoclonal Antibody
Pricing and availability is not currently available.