MilliporeSigma
  • Neuropeptides in the human celiac/superior mesenteric ganglionic complex: an immunohistochemical study.
Neuropeptides in the human celiac/superior mesenteric ganglionic complex: an immunohistochemical study.
Journal of chemical neuroanatomy (1993-03-01)
M Quartu, J M Polak, M Del Fiacco
ABSTRACT
The occurrence of vasoactive intestinal polypeptide (VIP), peptide histidine-isoleucine (PHI), calcitonin gene-related peptide (CGRP), substance P (SP), somatostatin (SOM), galanin (GAL) and enkephalins (ENK) is studied in the human celiac/superior mesenteric ganglionic complex of pre- and full-term newborns, and adult subjects by means of immunohistochemistry. The antisera used labelled nerve fibres and terminal-like networks for each examined peptide, as well as VIP- and SOM-positive postganglionic neurons. Differences in the relative amount and density of the structures immunoreactive to the various peptides were observed. Moreover, variations in the amount and type of labelled elements were appreciable for each peptide when specimens from subjects at perinatal and adult ages were compared. Double-labelling immunofluorescence for SP and each other peptide showed that co-localization with SP is very frequent for CGRP, moderate to scarce for GAL and SOM, and rare to absent for PHI, VIP and ENK. VIP-, ENK- and CGRP-immunolabeled perikarya bearing the morphological features of the small intensely fluorescent (SIF) cells occurred in the organ. The presence of a paraganglion in one of the specimens examined allowed the detection of VIP- and ENK-positive cell bodies and VIP-, ENK-, SP- and GAL-like immunoreactive varicose nerve fibres in it. The results obtained provide substantial morphological data in support of the involvement of the examined peptides in the chemical interneuronal signalling in the human celiac/superior mesenteric ganglia.