MilliporeSigma
  • Cloning, expression, purification, and biological activity of recombinant native and variant human alpha 1-antichymotrypsins.
Cloning, expression, purification, and biological activity of recombinant native and variant human alpha 1-antichymotrypsins.
The Journal of biological chemistry (1990-01-15)
H Rubin, Z M Wang, E B Nickbarg, S McLarney, N Naidoo, O L Schoenberger, J L Johnson, B S Cooperman
ABSTRACT
Human alpha 1-antichymotrypsin has been cloned, sequenced and expressed in Escherichia coli and recombinant protein as well as point-specific mutants have been purified and characterized. The corrected gene-deduced amino acid sequence has 45% overall identity with alpha 1-protease inhibitor, which is higher than the 42% previously reported (Chandra, T., Stackhouse, R., Kidd, V. J., Robson, J. H., and Woo, S. L. C. (1983) Biochemistry 22, 5055-5060). Recombinant antichymotrypsin (rACT) is similar to natural antichymotrypsin with respect to the specificity of its interactions with proteases. Its second-order rate constant for association with bovine chymotrypsin is 6-8 x 10(5) M-1 s-1, which is identical to that of the serum-derived inhibitor. Site-specific mutagenesis has been used to produce two variants of rACT in which the P1 position has been changed from leucine to either methionine (L358M-rACT) or arginine (L358R-rACT). L358M-rACT has a specificity of inhibitory activity toward serine proteases closely similar to that of native rACT. By contrast, the specificity of L358R-rACT is quite different from that of native rACT, most notably in efficiently inhibiting trypsin and human thrombin while showing a decreased ability to inhibit chymotrypsin.