MilliporeSigma
Studies on oxidative modifications of nucleic acid pyrimidine bases.
Nucleic acids symposium series (1984-01-01)
T Harayama, R Yanada, K Kotoji, F Yoneda, T Taga, K Osaki, T Nagamatsu
ABSTRACT
Oxidation of diacetylthymidine (4), triacetyluridine (5), and 5-fluoro-1,3-dimethyluracil (3) with m-chloroperbenzoic acid and cross-linkage of the bromohydrin (6) with the benzylamine and glycine ethyl ester are described. A Plausible mechanism for formation of oxidation products is also presented.