MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Characterization of nitrogen-rich biomaterial-derived biochars and their sorption for aromatic compounds.
Characterization of nitrogen-rich biomaterial-derived biochars and their sorption for aromatic compounds.
Environmental pollution (Barking, Essex : 1987) (2014-09-10)
Meng Zhang, Liang Shu, Xiaofang Shen, Xiaoying Guo, Shu Tao, Baoshan Xing, Xilong Wang
ABSTRACT
Biochars from nitrogen-rich biomaterials (i.e., α-amylase, chitin and zein) were produced at different temperatures (i.e. 170, 250, 350 and 450 °C) and characterized, and their sorption for phenanthrene, naphthalene and 1-naphthol was investigated. The organic carbon content normalized-sorption coefficient (Koc) of the tested compounds by biochars increased with increasing charring temperature, attributed to the reduction of O-containing polar moieties especially the O-alkyl components, and the newly created aromatic carbon domains. The N-heterocyclic ring structure formed during charring process may enhance π-π interactions between aromatics and the aromatic components in the resulting biochars. However, π-π interactions did not dominate sorption of aromatics by N-rich biochars. Sorption of the tested compounds by N-rich biochars was predominantly controlled by the hydrophobic interactions between these chemicals and the aromatic components in biochars. Both N- and O-containing polar moieties at the biochar surfaces negatively affected their sorption for aromatics.
Product Description
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Supelco
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
1-Naphthol, SAJ special grade, ≥99.0%
Supelco
Phenanthrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Carbon, rod, 150mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 12.5x12.5mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon, rod, 100mm, diameter 3.0mm, graphite, 100%
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Millipore
Activated charcoal, suitable for GC
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular