MilliporeSigma
  • Zoogloea oleivorans sp. nov., a floc-forming, petroleum hydrocarbon-degrading bacterium isolated from biofilm.
Zoogloea oleivorans sp. nov., a floc-forming, petroleum hydrocarbon-degrading bacterium isolated from biofilm.
International journal of systematic and evolutionary microbiology (2014-10-25)
Milán Farkas, András Táncsics, Balázs Kriszt, Tibor Benedek, Erika M Tóth, Zsuzsa Kéki, Péter G Veres, Sándor Szoboszlay
ABSTRACT
A floc-forming, Gram-stain-negative, petroleum hydrocarbon-degrading bacterial strain, designated Buc(T), was isolated from a petroleum hydrocarbon-contaminated site in Hungary. Phylogenetic analysis based on 16S rRNA gene sequences revealed that strain Buc(T) formed a distinct phyletic lineage within the genus Zoogloea. Its closest relative was found to be Zoogloea caeni EMB43(T) (97.2% 16S rRNA gene sequence similarity) followed by Zoogloea oryzae A-7(T) (95.9%), Zoogloea ramigera ATCC 19544(T) (95.5%) and Zoogloea resiniphila DhA-35(T) (95.4%). The level of DNA-DNA relatedness between strain Buc(T) and Z. caeni EMB43(T) was 31.6%. Cells of strain Buc(T) are facultatively aerobic, rod-shaped, and motile by means of a polar flagellum. The strain grew at temperatures of 5-35 °C (optimum 25-28 °C), and at pH 6.0-9.0 (optimum 6.5-7.5). The predominant fatty acids were C16:0, C10 : 0 3-OH, C12:0 and summed feature 3 (C16 : 1ω7c and/or iso-C15 : 0 2-OH). The major respiratory quinone was ubiquinone-8 (Q-8) and the predominant polar lipid was phosphatidylethanolamine. The genomic DNA G+C content was 63.2 mol%. On the basis of the chemotaxonomic, molecular and phenotypic data, isolate Buc(T) is considered to represent a novel species of the genus Zoogloea, for which the name Zoogloea oleivorans sp. nov. is proposed. The type strain is Buc(T) ( =DSM 28387(T) =NCAIM B 02570(T)).
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder