MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Twelve or 30 months of dual antiplatelet therapy after drug-eluting stents.
Twelve or 30 months of dual antiplatelet therapy after drug-eluting stents.
The New England journal of medicine (2014-11-18)
Laura Mauri, Dean J Kereiakes, Robert W Yeh, Priscilla Driscoll-Shempp, Donald E Cutlip, P Gabriel Steg, Sharon-Lise T Normand, Eugene Braunwald, Stephen D Wiviott, David J Cohen, David R Holmes, Mitchell W Krucoff, James Hermiller, Harold L Dauerman, Daniel I Simon, David E Kandzari, Kirk N Garratt, David P Lee, Thomas K Pow, Peter Ver Lee, Michael J Rinaldi, Joseph M Massaro
ABSTRACT
Dual antiplatelet therapy is recommended after coronary stenting to prevent thrombotic complications, yet the benefits and risks of treatment beyond 1 year are uncertain. Patients were enrolled after they had undergone a coronary stent procedure in which a drug-eluting stent was placed. After 12 months of treatment with a thienopyridine drug (clopidogrel or prasugrel) and aspirin, patients were randomly assigned to continue receiving thienopyridine treatment or to receive placebo for another 18 months; all patients continued receiving aspirin. The coprimary efficacy end points were stent thrombosis and major adverse cardiovascular and cerebrovascular events (a composite of death, myocardial infarction, or stroke) during the period from 12 to 30 months. The primary safety end point was moderate or severe bleeding. A total of 9961 patients were randomly assigned to continue thienopyridine treatment or to receive placebo. Continued treatment with thienopyridine, as compared with placebo, reduced the rates of stent thrombosis (0.4% vs. 1.4%; hazard ratio, 0.29 [95% confidence interval {CI}, 0.17 to 0.48]; P<0.001) and major adverse cardiovascular and cerebrovascular events (4.3% vs. 5.9%; hazard ratio, 0.71 [95% CI, 0.59 to 0.85]; P<0.001). The rate of myocardial infarction was lower with thienopyridine treatment than with placebo (2.1% vs. 4.1%; hazard ratio, 0.47; P<0.001). The rate of death from any cause was 2.0% in the group that continued thienopyridine therapy and 1.5% in the placebo group (hazard ratio, 1.36 [95% CI, 1.00 to 1.85]; P=0.05). The rate of moderate or severe bleeding was increased with continued thienopyridine treatment (2.5% vs. 1.6%, P=0.001). An elevated risk of stent thrombosis and myocardial infarction was observed in both groups during the 3 months after discontinuation of thienopyridine treatment. Dual antiplatelet therapy beyond 1 year after placement of a drug-eluting stent, as compared with aspirin therapy alone, significantly reduced the risks of stent thrombosis and major adverse cardiovascular and cerebrovascular events but was associated with an increased risk of bleeding. (Funded by a consortium of eight device and drug manufacturers and others; DAPT ClinicalTrials.gov number, NCT00977938.).
Product Description
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Ticlopidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Supelco
Ticlopidine hydrochloride, analytical standard, for drug analysis
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard