MilliporeSigma
  • Refinement of variant selection for the LDL cholesterol genetic risk score in the diagnosis of the polygenic form of clinical familial hypercholesterolemia and replication in samples from 6 countries.
Refinement of variant selection for the LDL cholesterol genetic risk score in the diagnosis of the polygenic form of clinical familial hypercholesterolemia and replication in samples from 6 countries.
Clinical chemistry (2014-11-22)
Marta Futema, Sonia Shah, Jackie A Cooper, KaWah Li, Ros A Whittall, Mahtab Sharifi, Olivia Goldberg, Euridiki Drogari, Vasiliki Mollaki, Albert Wiegman, Joep Defesche, Maria N D'Agostino, Antonietta D'Angelo, Paolo Rubba, Giuliana Fortunato, Małgorzata Waluś-Miarka, Robert A Hegele, Mary Aderayo Bamimore, Ronen Durst, Eran Leitersdorf, Monique T Mulder, Jeanine E Roeters van Lennep, Eric J G Sijbrands, John C Whittaker, Philippa J Talmud, Steve E Humphries
ABSTRACT
Familial hypercholesterolemia (FH) is an autosomal-dominant disorder caused by mutations in 1 of 3 genes. In the 60% of patients who are mutation negative, we have recently shown that the clinical phenotype can be associated with an accumulation of common small-effect LDL cholesterol (LDL-C)-raising alleles by use of a 12-single nucleotide polymorphism (12-SNP) score. The aims of the study were to improve the selection of SNPs and replicate the results in additional samples. We used ROC curves to determine the optimum number of LDL-C SNPs. For replication analysis, we genotyped patients with a clinical diagnosis of FH from 6 countries for 6 LDL-C-associated alleles. We compared the weighted SNP score among patients with no confirmed mutation (FH/M-), those with a mutation (FH/M+), and controls from a UK population sample (WHII). Increasing the number of SNPs to 33 did not improve the ability of the score to discriminate between FH/M- and controls, whereas sequential removal of SNPs with smaller effects/lower frequency showed that a weighted score of 6 SNPs performed as well as the 12-SNP score. Metaanalysis of the weighted 6-SNP score, on the basis of polymorphisms in CELSR2 (cadherin, EGF LAG 7-pass G-type receptor 2), APOB (apolipoprotein B), ABCG5/8 [ATP-binding cassette, sub-family G (WHITE), member 5/8], LDLR (low density lipoprotein receptor), and APOE (apolipoprotein E) loci, in the independent FH/M- cohorts showed a consistently higher score in comparison to the WHII population (P < 2.2 × 10(-16)). Modeling in individuals with a 6-SNP score in the top three-fourths of the score distribution indicated a >95% likelihood of a polygenic explanation of their increased LDL-C. A 6-SNP LDL-C score consistently distinguishes FH/M- patients from healthy individuals. The hypercholesterolemia in 88% of mutation-negative patients is likely to have a polygenic basis.