MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Contribution of volatiles to the antifungal effect of Lactobacillus paracasei in defined medium and yogurt.
Contribution of volatiles to the antifungal effect of Lactobacillus paracasei in defined medium and yogurt.
International journal of food microbiology (2014-12-03)
S D Aunsbjerg, A H Honoré, J Marcussen, P Ebrahimi, F K Vogensen, C Benfeldt, T Skov, S Knøchel
ABSTRACT
Lactic acid bacteria with antifungal properties can be used to control spoilage of food and feed. Previously, most of the identified metabolites have been isolated from cell-free fermentate of lactic acid bacteria with methods suboptimal for detecting possible contribution from volatiles to the antifungal activity. The role of volatile compounds in the antifungal activity of Lactobacillus paracasei DGCC 2132 in a chemically defined interaction medium (CDIM) and yogurt was therefore investigated with a sampling technique minimizing volatile loss. Diacetyl was identified as the major volatile produced by L. paracasei DGCC 2132 in CDIM. When the strain was added to a yogurt medium diacetyl as well as other volatiles also increased but the metabolome was more complex. Removal of L. paracasei DGCC 2132 cells from CDIM fermentate resulted in loss of both volatiles, including diacetyl, and the antifungal activity towards two strains of Penicillium spp. When adding diacetyl to CDIM or yogurt without L. paracasei DGCC 2132, marked inhibition was observed. Besides diacetyl, the antifungal properties of acetoin were examined, but no antifungal activity was observed. Overall, the results demonstrate the contribution of diacetyl in the antifungal effect of L. paracasei DGCC 2132 and indicate that the importance of volatiles may have been previously underestimated.
Product Description
Supelco
2,3-Butanedione, analytical standard
Sigma-Aldrich
Acetoin, primarily dimer, ≥95%, FG
Sigma-Aldrich
Acetoin, natural, ≥95%, FG
Sigma-Aldrich
2,3-Butanedione, 97%
Sigma-Aldrich
Acetoin, May exist as crystalline dimer
Supelco
Acetoin, analytical standard